KEY432441856 Key Organics / BIONET

3-(4-chlorobenzenesulfonyl)-6-fluoro-1,4-dihydroquinolin-4-one

  • MDL number MFCD05900155
  • Cas Number : 866727-75-5
  • Molecular Weight : 337.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442030

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3494_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart