KEY432441857 Key Organics / BIONET

4-[(2-hydrazinylpyridin-3-yl)sulfonyl]morpholine

  • MDL number MFCD12174508
  • Cas Number : 74186-30-4
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(2-hydrazino-3-pyridinyl)sulfonyl]morpholine
Inchi Code 1S/C9H14N4O3S/c10-12-9-8(2-1-3-11-9)17(14,15)13-4-6-16-7-5-13/h1-3H,4-7,10H2,(H,11,12)
Inchi Key XOMYEJOXHUVXBJ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442031

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3495_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart