KEY432441862 Key Organics / BIONET

methyl 5-methyl-1-(4-methylphenyl)-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD26130115
  • Cas Number : 1638612-45-9
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-methyl-1-(p-tolyl)-1H-1,2,4-triazole-3-carboxylate
Inchi Code 1S/C12H13N3O2/c1-8-4-6-10(7-5-8)15-9(2)13-11(14-15)12(16)17-3/h4-7H,1-3H3
Inchi Key WYFLNXJUSJRQNO-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442036

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3500_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart