KEY432441870 Key Organics / BIONET

ethyl 3-amino-1,1-dioxo-1lambda6-benzothiophene-2-carboxylate

  • MDL number MFCD09743336
  • Cas Number : 936074-60-1
  • Molecular Weight : 253.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442044

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3508_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart