KEY432441872 Key Organics / BIONET

(Z)-2-[2-(2-ethoxyphenyl)-5-methyl-1,3-oxazol-4-yl]-N'-hydroxyethanimidamide

  • MDL number MFCD26130242
  • Cas Number : 1638617-32-9
  • Molecular Weight : 275.308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3510_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart