KEY432441874 Key Organics / BIONET

6-[(4-methylpiperidin-1-yl)sulfonyl]-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one

  • MDL number MFCD18788421
  • Cas Number : 1291834-57-5
  • Molecular Weight : 296.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442048

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3512_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart