KEY432441877 Key Organics / BIONET

1-(3-fluorophenyl)-4-methoxy-6-oxo-1,6-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD26130258
  • Cas Number : 1638612-42-6
  • Molecular Weight : 264.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442051

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3515_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart