KEY432441880 Key Organics / BIONET

4-(4-methylphenyl)-1,1-dioxo-4H-1lambda6,4-benzothiazine-2-carbonitrile

  • MDL number MFCD18788900
  • Cas Number : 1291832-37-5
  • Molecular Weight : 296.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(p-tolyl)-4H-benzo[b][1,4]thiazine-2-carbonitrile 1,1-dioxide
Inchi Code 1S/C16H12N2O2S/c1-12-6-8-13(9-7-12)18-11-14(10-17)21(19,20)16-5-3-2-4-15(16)18/h2-9,11H,1H3
Inchi Key SBXSVYJWWUHTEP-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442054

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3518_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart