KEY432441881 Key Organics / BIONET

3-(N-methyl4-chlorobenzenesulfonamido)thiophene-2-carboxylic acid

  • MDL number MFCD26130279
  • Cas Number : 1638612-62-0
  • Molecular Weight : 331.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442055

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3519_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart