KEY432441882 Key Organics / BIONET

ethyl 2-ethyl-5-methyl-1,1-dioxo-2H-1lambda6,2,6-thiadiazine-4-carboxylate

  • MDL number MFCD27623217
  • Cas Number : 1340702-75-1
  • Molecular Weight : 246.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442056

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3520_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart