KEY432441885 Key Organics / BIONET

3-(1H-imidazole-1-carbonyl)-1H-indazole

  • MDL number MFCD26130293
  • Cas Number : 1025910-84-2
  • Molecular Weight : 212.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442059

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3523_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart