KEY432441886 Key Organics / BIONET

1-(2H-1,3-benzodioxole-5-carbonyl)-1H-imidazole

  • MDL number MFCD17292452
  • Cas Number : 135696-67-2
  • Molecular Weight : 216.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442060

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3524_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart