KEY432441888 Key Organics / BIONET

1-{[1,2,4]triazolo[1,5-a]pyrimidine-2-carbonyl}-1H-imidazole

  • MDL number MFCD26130294
  • Cas Number : 1638612-65-3
  • Molecular Weight : 214.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442062

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3526_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart