KEY432441889 Key Organics / BIONET

3-(1H-imidazole-1-carbonyl)-1-methyl-1H-indazole

  • MDL number MFCD26130296
  • Cas Number : 148672-80-4
  • Molecular Weight : 226.239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442063

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3527_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart