KEY432441890 Key Organics / BIONET

methyl 5-(chlorosulfonyl)-3-ethyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD26129973
  • Molecular Weight : 252.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3528_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart