KEY432441891 Key Organics / BIONET

ethyl 3-(2H-1,3-benzodioxol-5-yl)-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD06618106
  • Cas Number : 884982-97-2
  • Molecular Weight : 262.221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442065

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3529_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart