KEY432441897 Key Organics / BIONET

3-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]pyridine-2-thiol

  • MDL number MFCD19104981
  • Cas Number : 1325306-72-6
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442071

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3536_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart