KEY432441900 Key Organics / BIONET

1'-methyl-3',4'-dihydro-1'H-spiro[piperidine-4,2'-quinazoline]-4'-one

  • MDL number MFCD02929845
  • Cas Number : 438621-93-3
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442074

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3539_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart