KEY432441906 Key Organics / BIONET

5-(2,4-dioxo-1,2,3,4-tetrahydroquinazolin-3-yl)pentanoic acid

  • MDL number MFCD05027538
  • Cas Number : 571172-12-8
  • Molecular Weight : 262.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442080

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3546_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart