KEY432441908 Key Organics / BIONET

ethyl 6-(chloromethyl)-4-(2,3-dimethoxyphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD02374294
  • Cas Number : 380589-29-7
  • Molecular Weight : 354.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442082

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3548_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart