KEY432441909 Key Organics / BIONET

2-(trifluoromethyl)imidazo[1,2-a]pyrimidine

  • MDL number MFCD19705789
  • Cas Number : 73221-25-7
  • Molecular Weight : 187.125

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442083

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3549_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart