KEY432441915 Key Organics / BIONET

(Z)-2-(4-fluorophenyl)-N'-hydroxyethanimidamide

  • MDL number MFCD07606939
  • Cas Number : 925252-30-8
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442089

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3555_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart