KEY432441918 Key Organics / BIONET

3-(N-methyl4-tert-butylbenzenesulfonamido)thiophene-2-carboxylic acid

  • MDL number MFCD19104880
  • Cas Number : 1111300-43-6
  • Molecular Weight : 353.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442092

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3558_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart