KEY432441922 Key Organics / BIONET

3-(3-chloro-4-methylbenzenesulfonamido)thiophene-2-carboxylic acid

  • MDL number MFCD19105019
  • Cas Number : 1325304-03-7
  • Molecular Weight : 331.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442096

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3562_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart