KEY432441923 Key Organics / BIONET

3-(4-propylbenzenesulfonamido)thiophene-2-carboxylic acid

  • MDL number MFCD13696880
  • Cas Number : 1153238-87-9
  • Molecular Weight : 325.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442097

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3563_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart