KEY432441924 Key Organics / BIONET

3-(N-methyl4-bromobenzenesulfonamido)thiophene-2-carboxylic acid

  • MDL number MFCD18804084
  • Cas Number : 1291486-57-1
  • Molecular Weight : 376.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442098

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3564_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart