KEY432441930 Key Organics / BIONET

1-[3-(1H-pyrrol-1-yl)thieno[2,3-b]pyridine-2-carbonyl]-1H-imidazole

  • MDL number MFCD19105030
  • Cas Number : 1325305-13-2
  • Molecular Weight : 294.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-imidazol-1-ylcarbonyl)-3-(1H-pyrrol-1-yl)thieno[2,3-b]pyridine
Inchi Code 1S/C15H10N4OS/c20-15(19-9-6-16-10-19)13-12(18-7-1-2-8-18)11-4-3-5-17-14(11)21-13/h1-10H
Inchi Key JKDRPAAKYDVDFL-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442104

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3570_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart