KEY432441932 Key Organics / BIONET

6-fluoro-1-methyl-4-oxo-7-(piperidin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD14729054
  • Cas Number : 1030120-88-7
  • Molecular Weight : 304.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3572_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart