KEY432441939 Key Organics / BIONET

6-methyl-3-(propan-2-yl)-2-sulfanylidene-1H,2H,3H,4H,5H-pyrrolo[3,2-d]pyrimidin-4-one

  • MDL number MFCD18915339
  • Cas Number : 1325303-62-5
  • Molecular Weight : 223.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442113

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3579_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart