KEY432441945 Key Organics / BIONET

1-[2-(2H-1,3-benzodioxol-5-yl)-2-hydroxyethyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD25952438
  • Cas Number : 1338691-87-4
  • Molecular Weight : 277.236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3585_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart