KEY432441950 Key Organics / BIONET

2,4-diethyl 1-(2-oxo-2-phenylethyl)-1H-imidazole-2,4-dicarboxylate

  • MDL number MFCD28155226
  • Cas Number : 1638612-75-5
  • Molecular Weight : 330.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442124

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3590_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart