KEY432441952 Key Organics / BIONET

4-chloro-6-{4-[3-(pyridin-3-yl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl}pyrimidine

  • MDL number MFCD21640442
  • Cas Number : 1516585-08-2
  • Molecular Weight : 342.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442126

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3592_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart