KEY432441956 Key Organics / BIONET

{[3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD28155248
  • Cas Number : 1638612-54-0
  • Molecular Weight : 260.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442130

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3596_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart