KEY432441964 Key Organics / BIONET

[3-(thiophen-2-yl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD06202709
  • Cas Number : 194546-14-0
  • Molecular Weight : 180.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [3-(2-thienyl)-1H-pyrazol-4-yl]methanol
Inchi Code 1S/C8H8N2OS/c11-5-6-4-9-10-8(6)7-2-1-3-12-7/h1-4,11H,5H2,(H,9,10)
Inchi Key FTMOMXZFKPMKAI-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442138

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3604_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart