KEY432441967 Key Organics / BIONET

5-(methoxymethyl)-2-(piperazin-1-yl)-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one hydrochloride

  • MDL number MFCD28155233
  • Cas Number : 1638612-79-9
  • Molecular Weight : 300.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442141

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3607_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart