KEY432441970 Key Organics / BIONET

2-(3-aminopyrrolidine-1-carbonyl)-6-methyl-4H,5H-pyrazolo[1,5-a]pyrazin-4-one hydrochloride

  • MDL number MFCD28155235
  • Cas Number : 1638612-67-5
  • Molecular Weight : 297.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-aminopyrrolidine-1-carbonyl)-6-methylpyrazolo[1,5-a]pyrazin-4(5H)-one hydrochloride
Inchi Code 1S/C12H15N5O2.ClH/c1-7-5-17-10(11(18)14-7)4-9(15-17)12(19)16-3-2-8(13)6-16;/h4-5,8H,2-3,6,13H2,1H3,(H,14,18);1H
Inchi Key LUZSAQCSNJXQFJ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442144

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3610_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart