KEY432441972 Key Organics / BIONET

4-(4-fluorophenyl)-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD25953218
  • Cas Number : 862112-00-3
  • Molecular Weight : 207.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442146

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3612_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart