KEY432441975 Key Organics / BIONET

1-(2-methoxyphenyl)-5-(pyridin-4-yl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD14703448
  • Cas Number : 1223881-43-3
  • Molecular Weight : 296.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442149

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3615_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart