KEY432441978 Key Organics / BIONET

1-(2-chlorophenyl)-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11588447
  • Cas Number : 1006597-09-6
  • Molecular Weight : 246.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442152

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3618_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart