KEY432441980 Key Organics / BIONET

3-(1H-pyrrol-3-yl)-1H-pyrazole-5-carbohydrazide

  • MDL number MFCD28155239
  • Molecular Weight : 191.194

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442154

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3620_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart