KEY432441981 Key Organics / BIONET

tert-butyl 8-methoxy-1H,2H,3H,4H,5H-pyrido[4,3-b]indole-2-carboxylate

  • MDL number MFCD24471146
  • Cas Number : 1186099-85-3
  • Molecular Weight : 302.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442155

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart