KEY432441986 Key Organics / BIONET

4-{[(thiophen-2-yl)methyl]amino}-1,2-dihydroquinazolin-2-one

  • MDL number MFCD14729149
  • Cas Number : 1031970-30-5
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442160

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3626_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart