KEY432441993 Key Organics / BIONET

8-chloro-3-methyl-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD08272087
  • Cas Number : 68774-78-7
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-chloro-3-methyl[1,2,4]triazolo[4,3-a]pyrazine
Inchi Code 1S/C6H5ClN4/c1-4-9-10-6-5(7)8-2-3-11(4)6/h2-3H,1H3
Inchi Key RIWAMVRPNJEQJK-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442167

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3633_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart