KEY432441997 Key Organics / BIONET

methyl 4-oxo-2-sulfanylidene-1,2,3,4-tetrahydroquinazoline-7-carboxylate

  • MDL number MFCD04063080
  • Cas Number : 422277-15-4
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442171

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3637_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart