KEY432442004 Key Organics / BIONET

6-fluoro-3-(piperidin-4-yl)-1,2,3,4-tetrahydroquinazoline-2,4-dione hydrochloride

  • MDL number MFCD28155257
  • Cas Number : 1638612-46-0
  • Molecular Weight : 299.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442178

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3644_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart