KEY432442006 Key Organics / BIONET

1-benzyl-5-(pyridin-4-yl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD25950189
  • Cas Number : 1338688-22-4
  • Molecular Weight : 280.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-5-(4-pyridinyl)-1H-1,2,3-triazole-4-carboxylic acid
Inchi Code 1S/C15H12N4O2/c20-15(21)13-14(12-6-8-16-9-7-12)19(18-17-13)10-11-4-2-1-3-5-11/h1-9H,10H2,(H,20,21)
Inchi Key NMASXQCMUBAFJG-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442180

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3646_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart