KEY432442008 Key Organics / BIONET

2-phenyl-1H,2H,3H-pyrazolo[1,5-a]imidazole-7-carboxylic acid

  • MDL number MFCD27974511
  • Cas Number : 946150-84-1
  • Molecular Weight : 229.239

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442182

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3648_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart