KEY432442017 Key Organics / BIONET

2-chloro-N-(2-methoxy-5-nitrophenyl)acetamide

  • MDL number MFCD02580945
  • Cas Number : 58795-63-4
  • Molecular Weight : 244.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-2993_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart