KEY432442023 Key Organics / BIONET

(1S,3S)-3-aminocyclohexan-1-ol hydrochloride

  • MDL number MFCD22689398
  • Cas Number : 721884-81-7
  • Molecular Weight : 151.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442216

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4867_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart