KEY432442025 Key Organics / BIONET

(1R,3R)-3-aminocyclohexan-1-ol hydrochloride

  • MDL number MFCD26954737
  • Cas Number : 124555-43-7
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442218

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4869_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart